siRNA Details
| Gene Symbol: Tpr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107185.1 | NP_001100655.2 | 30, 31 | 4319 | ||
| XM_006249972.1 | 30, 31 | 4342 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tpr| Gene Symbol: Tpr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107185.1 | NP_001100655.2 | 30, 31 | 4319 | ||
| XM_006249972.1 | 30, 31 | 4342 | ||||