siRNA Details
| Gene Symbol: Tpr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107185.1 | NP_001100655.2 | 49, 50 | 7107 | ||
| XM_006249972.1 | 49, 50 | 7130 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tpr| Gene Symbol: Tpr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107185.1 | NP_001100655.2 | 49, 50 | 7107 | ||
| XM_006249972.1 | 49, 50 | 7130 | ||||