siRNA Details
| Gene Symbol: Tor3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009683.1 | NP_001009683.1 | 3 | 549 | ||
| GenBank mRNA | BC087678.1 | - | 3 | 549 | 7318816 | |
| FQ226439.1 | - | 1 | 110 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tor3a| Gene Symbol: Tor3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009683.1 | NP_001009683.1 | 3 | 549 | ||
| GenBank mRNA | BC087678.1 | - | 3 | 549 | 7318816 | |
| FQ226439.1 | - | 1 | 110 | |||