siRNA Details
| Gene Symbol: Astn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170603.1 | NP_001164074.1 | 20 | 3623 | ||
| XM_008769640.1 | XP_008767862.1 | 17 | 3041 | |||
| XM_008769641.1 | XP_008767863.1 | 17 | 3016 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Astn1| Gene Symbol: Astn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170603.1 | NP_001164074.1 | 20 | 3623 | ||
| XM_008769640.1 | XP_008767862.1 | 17 | 3041 | |||
| XM_008769641.1 | XP_008767863.1 | 17 | 3016 | |||