siRNA Details
| Gene Symbol: Tnn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107189.2 | NP_001100659.2 | 15 | 3752 | ||
| XM_008769642.1 | XP_008767864.1 | 16 | 4149 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tnn| Gene Symbol: Tnn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107189.2 | NP_001100659.2 | 15 | 3752 | ||
| XM_008769642.1 | XP_008767864.1 | 16 | 4149 | |||