siRNA Details
| Gene Symbol: Rc3h1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769698.1 | XP_008767920.1 | 9 | 1632 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rc3h1| Gene Symbol: Rc3h1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008769698.1 | XP_008767920.1 | 9 | 1632 | ||