siRNA Details
| Gene Symbol: Serpinc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012027.1 | NP_001012027.1 | 4 | 795 | ||
| XM_008769647.1 | XP_008767869.1 | 4 | 740 | |||
| GenBank mRNA | AY321346.1 | - | 654 | |||
| BC088467.1 | - | 4 | 795 | 7127992 | ||
| FQ210275.1 | - | 4 | 779 | |||
| FQ210716.1 | - | 4 | 815 | |||
| FQ210762.1 | - | 4 | 813 | |||
| FQ211135.1 | - | 4 | 813 | |||
| FQ219160.1 | - | 4 | 814 | |||
| FQ219233.1 | - | 4 | 778 | |||

Serpinc1
Genomic Map