siRNA Details
| Gene Symbol: Dars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034143.1 | NP_001029315.1 | 16, 17 | 1858 | ||
| GenBank mRNA | BC105774.1 | - | 16, 17 | 1858 | 7930977 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dars2| Gene Symbol: Dars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034143.1 | NP_001029315.1 | 16, 17 | 1858 | ||
| GenBank mRNA | BC105774.1 | - | 16, 17 | 1858 | 7930977 | |