siRNA Details
| Gene Symbol: Gorab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100563.1 | NP_001094033.1 | 2 | 241 | ||
| XM_008769650.1 | XP_008767872.1 | 1 | 817 | |||
| GenBank mRNA | BC160825.1 | - | 2 | 241 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gorab| Gene Symbol: Gorab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100563.1 | NP_001094033.1 | 2 | 241 | ||
| XM_008769650.1 | XP_008767872.1 | 1 | 817 | |||
| GenBank mRNA | BC160825.1 | - | 2 | 241 | ||