siRNA Details
| Gene Symbol: Gorab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100563.1 | NP_001094033.1 | 5 | 1093 | ||
| XM_008769650.1 | XP_008767872.1 | 4 | 1669 | |||
| GenBank mRNA | BC086370.1 | - | 3 | 572 | ||
| BC160825.1 | - | 5 | 1093 | |||

Gorab
Genomic Map