siRNA Details
| Gene Symbol: Atf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107196.1 | NP_001100666.1 | 13 | 1610 | ||
| XM_008769738.1 | XP_008767960.1 | 13 | 1666 | |||
| GenBank mRNA | BC168890.1 | - | 13 | 1593 | 7108280 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atf6| Gene Symbol: Atf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107196.1 | NP_001100666.1 | 13 | 1610 | ||
| XM_008769738.1 | XP_008767960.1 | 13 | 1666 | |||
| GenBank mRNA | BC168890.1 | - | 13 | 1593 | 7108280 | |