siRNA Details
Gene Symbol: Tomm40l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083338.1 | NP_001076807.1 | 5 | 459 | ||
XM_006250247.2 | XP_006250309.1 | 6 | 938 | |||
GenBank mRNA | AB274732.1 | - | 5 | 459 |
