siRNA Details
| Gene Symbol: Brox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025669.1 | NP_001020840.1 | 12 | 1187 | ||
| XM_006250354.2 | XP_006250416.1 | 10 | 1037 | |||
| GenBank mRNA | BC097348.1 | - | 12 | 1187 | 7460070 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Brox| Gene Symbol: Brox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025669.1 | NP_001020840.1 | 12 | 1187 | ||
| XM_006250354.2 | XP_006250416.1 | 10 | 1037 | |||
| GenBank mRNA | BC097348.1 | - | 12 | 1187 | 7460070 | |