siRNA Details
| Gene Symbol: Dtl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_223074.7 | XP_223074.5 | 1, 2 | 217 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dtl| Gene Symbol: Dtl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_223074.7 | XP_223074.5 | 1, 2 | 217 | ||