siRNA Details
| Gene Symbol: Diexf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013986.2 | NP_001014008.1 | 11 | 1986 | ||
| XM_006250482.2 | XP_006250544.1 | 12 | 2176 | |||
| GenBank mRNA | BC087114.1 | - | 11 | 1940 | 7193786 | |
| FQ217506.1 | - | 12 | 2159 | |||

Diexf
Genomic Map