siRNA Details
| Gene Symbol: Diexf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013986.2 | NP_001014008.1 | 6 | 838 | ||
| XM_006250482.2 | XP_006250544.1 | 7 | 1028 | |||
| GenBank mRNA | BC087114.1 | - | 6 | 792 | 7193786 | |
| FQ217506.1 | - | 7 | 1011 | |||

Diexf
Genomic Map