siRNA Details
| Gene Symbol: Nudt9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006991.1 | NP_001006992.1 | 2, 3 | 451 | ||
| XM_006250630.2 | XP_006250692.1 | 1, 2 | 252 | |||
| XM_006250631.2 | XP_006250693.1 | 2, 3 | 274 | |||
| GenBank mRNA | BC083722.1 | - | 2, 3 | 451 | 7190984 | |

Nudt9
Genomic Map