siRNA Details
| Gene Symbol: Lin54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100564.1 | NP_001094034.1 | 4 | 1166 | ||
| XM_006250657.2 | XP_006250719.1 | 5 | 1282 | |||
| XM_006250658.2 | XP_006250720.1 | 5 | 1312 | |||
| XM_006250659.2 | XP_006250721.1 | 5 | 1477 | |||
| XM_006250661.2 | XP_006250723.1 | 5 | 649 | |||
| XM_008770022.1 | XP_008768244.1 | 5 | 619 | |||
| GenBank mRNA | BC082043.1 | - | 2 | 372 | ||
| BC099076.1 | - | 2 | 203 | |||

Lin54
Genomic Map