siRNA Details
Gene Symbol: Lin54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100564.1 | NP_001094034.1 | 2, 3 | 842 | ||
XM_006250657.2 | XP_006250719.1 | 3, 4 | 958 | |||
XM_006250658.2 | XP_006250720.1 | 3, 4 | 988 | |||
XM_006250659.2 | XP_006250721.1 | 3, 4 | 1153 | |||
XM_006250661.2 | XP_006250723.1 | 3, 4 | 325 | |||
XM_008770022.1 | XP_008768244.1 | 3, 4 | 295 | |||
GenBank mRNA | EV774847.1 | - | 2, 3 | 449 |
