siRNA Details
| Gene Symbol: Lin54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100564.1 | NP_001094034.1 | 5 | 1236 | ||
| XM_006250657.2 | XP_006250719.1 | 6 | 1352 | |||
| XM_006250658.2 | XP_006250720.1 | 6 | 1382 | |||
| XM_006250659.2 | XP_006250721.1 | 6 | 1547 | |||
| XM_006250661.2 | XP_006250723.1 | 6 | 719 | |||
| XM_008770022.1 | XP_008768244.1 | 6 | 689 | |||
| GenBank mRNA | BC082043.1 | - | 3 | 442 | ||
| BC099076.1 | - | 3 | 273 | |||

Lin54
Genomic Map