siRNA Details
| Gene Symbol: Rasgef1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002728083.4 | XP_002728129.2 | 6, 7 | 891 | ||
| XM_008770077.1 | XP_008768299.1 | 6, 7 | 888 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rasgef1b| Gene Symbol: Rasgef1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002728083.4 | XP_002728129.2 | 6, 7 | 891 | ||
| XM_008770077.1 | XP_008768299.1 | 6, 7 | 888 | |||