siRNA Details
| Gene Symbol: Paqr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012033.1 | NP_001012033.1 | 4 | 802 | ||
| XM_008770025.1 | XP_008768247.1 | 4 | 864 | |||
| GenBank mRNA | BC079418.1 | - | 4 | 802 | 7113653 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Paqr3| Gene Symbol: Paqr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012033.1 | NP_001012033.1 | 4 | 802 | ||
| XM_008770025.1 | XP_008768247.1 | 4 | 864 | |||
| GenBank mRNA | BC079418.1 | - | 4 | 802 | 7113653 | |