siRNA Details
Gene Symbol: Stbd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013988.1 | NP_001014010.1 | 2 | 809 | ||
GenBank mRNA | BC089867.1 | - | 2 | 809 | 7321620 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Stbd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013988.1 | NP_001014010.1 | 2 | 809 | ||
GenBank mRNA | BC089867.1 | - | 2 | 809 | 7321620 |