siRNA Details
| Gene Symbol: Cdkl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012035.1 | NP_001012035.1 | 10 | 1878 | ||
| XM_006250737.2 | XP_006250799.1 | 10 | 2006 | |||
| XM_006250738.2 | XP_006250800.1 | 11 | 2115 | |||
| XM_006250739.2 | XP_006250801.1 | 10 | 2506 | |||
| XM_006250740.2 | XP_006250802.1 | 11 | 2567 | |||
| XM_006250741.2 | XP_006250803.1 | 10 | 2458 | |||
| XM_008770028.1 | XP_008768250.1 | 11 | 2615 | |||
| XM_008770029.1 | XP_008768251.1 | 10 | 2462 | |||
| GenBank mRNA | AY383681.1 | - | 10 | 1571 | ||
| BC083590.1 | - | 10 | 1878 | 7132550 | ||

Cdkl2
Genomic Map