siRNA Details
| Gene Symbol: Grsf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100890.1 | NP_001094360.1 | 3 | 560 | ||
| XM_006250775.2 | XP_006250837.1 | 3 | 447 | |||
| GenBank mRNA | FM050032.1 | - | 3 | 521 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Grsf1| Gene Symbol: Grsf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100890.1 | NP_001094360.1 | 3 | 560 | ||
| XM_006250775.2 | XP_006250837.1 | 3 | 447 | |||
| GenBank mRNA | FM050032.1 | - | 3 | 521 | ||