siRNA Details
| Gene Symbol: Exoc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024770.1 | NP_001019941.1 | 6 | 813 | ||
| GenBank mRNA | BC097392.1 | - | 6 | 813 | 7453203 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Exoc1| Gene Symbol: Exoc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024770.1 | NP_001019941.1 | 6 | 813 | ||
| GenBank mRNA | BC097392.1 | - | 6 | 813 | 7453203 | |