siRNA Details
| Gene Symbol: Srd5a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013990.1 | NP_001014012.1 | 4 | 468 | ||
| XM_006250897.1 | XP_006250959.1 | 4 | 506 | |||
| GenBank mRNA | BC086584.1 | - | 4 | 468 | 7307843 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Srd5a3| Gene Symbol: Srd5a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013990.1 | NP_001014012.1 | 4 | 468 | ||
| XM_006250897.1 | XP_006250959.1 | 4 | 506 | |||
| GenBank mRNA | BC086584.1 | - | 4 | 468 | 7307843 | |