siRNA Details
| Gene Symbol: Slain2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107214.1 | NP_001100684.1 | 3 | 925 | ||
| XM_006250898.2 | XP_006250960.1 | 3 | 997 | |||
| XM_006250899.2 | XP_006250961.1 | 3 | 996 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slain2| Gene Symbol: Slain2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107214.1 | NP_001100684.1 | 3 | 925 | ||
| XM_006250898.2 | XP_006250960.1 | 3 | 997 | |||
| XM_006250899.2 | XP_006250961.1 | 3 | 996 | |||