siRNA Details
| Gene Symbol: Guf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107215.1 | NP_001100685.1 | 12 | 1546 | ||
| XM_008770207.1 | XP_008768429.1 | 12 | 1949 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Guf1| Gene Symbol: Guf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107215.1 | NP_001100685.1 | 12 | 1546 | ||
| XM_008770207.1 | XP_008768429.1 | 12 | 1949 | |||