siRNA Details
| Gene Symbol: Pds5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083624.1 | NP_001077093.1 | 21, 22 | 2509 | ||
| XM_008770153.1 | XP_008768375.1 | 21, 22 | 2543 | |||
| XM_008770154.1 | XP_008768376.1 | 20, 21 | 2456 | |||
| GenBank mRNA | EF460313.1 | - | 21, 22 | 2509 | ||

Pds5a
Genomic Map