siRNA Details
| Gene Symbol: Wdr19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191679.2 | NP_001178608.2 | 15 | 1570 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdr19| Gene Symbol: Wdr19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191679.2 | NP_001178608.2 | 15 | 1570 | ||