siRNA Details
| Gene Symbol: Arap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107216.1 | NP_001100686.1 | 2 | 223 | ||
| XM_008770184.1 | XP_008768406.1 | 2 | 234 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arap2| Gene Symbol: Arap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107216.1 | NP_001100686.1 | 2 | 223 | ||
| XM_008770184.1 | XP_008768406.1 | 2 | 234 | |||