siRNA Details
| Gene Symbol: Zbtb49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107223.1 | NP_001100693.1 | 3 | 1059 | ||
| XM_006251092.2 | XP_006251154.1 | 3 | 1085 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zbtb49| Gene Symbol: Zbtb49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107223.1 | NP_001100693.1 | 3 | 1059 | ||
| XM_006251092.2 | XP_006251154.1 | 3 | 1085 | |||