siRNA Details
| Gene Symbol: Dok7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130062.1 | NP_001123534.1 | 8 | 1594 | ||
| XM_006251239.2 | XP_006251301.1 | 9 | 2411 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dok7| Gene Symbol: Dok7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130062.1 | NP_001123534.1 | 8 | 1594 | ||
| XM_006251239.2 | XP_006251301.1 | 9 | 2411 | |||