siRNA Details
| Gene Symbol: Mfsd10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191680.1 | NP_001178609.1 | 12 | 1320 | ||
| XM_006251243.2 | XP_006251305.1 | 13 | 1622 | |||
| XM_006251244.2 | XP_006251306.1 | 13 | 1619 | |||
| XM_006251245.2 | XP_006251307.1 | 13 | 1455 | |||
| XM_006251246.2 | XP_006251308.1 | 13 | 1452 | |||

Mfsd10
Genomic Map