siRNA Details
| Gene Symbol: Sh3bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100684.1 | NP_001094154.1 | 2 | 232 | ||
| XM_006251247.2 | XP_006251309.1 | 2 | 1271 | |||
| XM_006251248.2 | XP_006251310.1 | 2 | 1109 | |||
| XM_006251249.2 | XP_006251311.1 | 2 | 1323 | |||
| XM_006251251.2 | XP_006251313.1 | 2 | 181 | |||
| XM_006251252.2 | XP_006251314.1 | 2 | 3809 | |||
| XM_006251253.2 | XP_006251315.1 | 2 | 1045 | |||
| XM_008770284.1 | XP_008768506.1 | 3 | 4776 | |||
| XM_008770285.1 | XP_008768507.1 | 2 | 438 | |||
| GenBank mRNA | CB762696.1 | - | 2 | 240 | ||
| FQ218289.1 | - | 2 | 180 | |||

Sh3bp2
Genomic Map