siRNA Details
Gene Symbol: LOC103690085 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006251409.2 | XP_006251471.1 | 5 | 2412 |
Gene Symbol: Fam193a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_223536.8 | XP_223536.5 | 18 | 3472 | ||
GenBank mRNA | FQ225369.1 | - | 1859 |
