siRNA Details
| Gene Symbol: Cfap99 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008770377.1 | XP_008768599.1 | 13, 14 | 1585 | ||
| XM_008770378.1 | XP_008768600.1 | 14, 15 | 1696 | |||
| XM_008770379.1 | XP_008768601.1 | 13, 14 | 1608 | |||
| XM_008770380.1 | XP_008768602.1 | 11, 12 | 5265 | |||
| XM_008770381.1 | XP_008768603.1 | 12, 13 | 2273 | |||

Cfap99
Genomic Map