siRNA Details
Gene Symbol: Eif4enif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107230.1 | NP_001100700.1 | 7, 8 | 1125 | ||
XM_006251290.2 | XP_006251352.1 | 7, 8 | 1257 | |||
XM_006251291.2 | XP_006251353.1 | 7, 8 | 1013 | |||
XM_008770296.1 | XP_008768518.1 | 6, 7 | 1720 | |||
GenBank mRNA | FQ212399.1 | - | 7, 8 | 1069 |
