siRNA Details
Gene Symbol: Eif4enif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107230.1 | NP_001100700.1 | 14, 15 | 2156 | ||
XM_006251290.2 | XP_006251352.1 | 14, 15 | 2288 | |||
XM_006251291.2 | XP_006251353.1 | 14, 15 | 2044 | |||
XM_008770296.1 | XP_008768518.1 | 13, 14 | 2751 | |||
GenBank mRNA | FQ212399.1 | - | 14, 15 | 2100 |
