siRNA Details
| Gene Symbol: Drg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009685.1 | NP_001009685.1 | 3 | 324 | ||
| GenBank mRNA | BC088410.1 | - | 3 | 324 | 7189810 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Drg1| Gene Symbol: Drg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009685.1 | NP_001009685.1 | 3 | 324 | ||
| GenBank mRNA | BC088410.1 | - | 3 | 324 | 7189810 | |