siRNA Details
| Gene Symbol: Pik3ip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017453.1 | NP_001017453.1 | 1 | 146 | ||
| XM_006251293.2 | XP_006251355.1 | 1 | 1292 | |||
| GenBank mRNA | BC092208.1 | - | 1 | 146 | 7314987 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pik3ip1| Gene Symbol: Pik3ip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017453.1 | NP_001017453.1 | 1 | 146 | ||
| XM_006251293.2 | XP_006251355.1 | 1 | 1292 | |||
| GenBank mRNA | BC092208.1 | - | 1 | 146 | 7314987 | |