siRNA Details
| Gene Symbol: Rnf215 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107234.1 | NP_001100704.1 | 3 | 565 | ||
| XM_006251300.1 | XP_006251362.1 | 3 | 591 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf215| Gene Symbol: Rnf215 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107234.1 | NP_001100704.1 | 3 | 565 | ||
| XM_006251300.1 | XP_006251362.1 | 3 | 591 | |||