siRNA Details
Gene Symbol: Urgcp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077660.2 | NP_001071128.1 | 1, 2 | 51 | ||
XM_006251313.2 | XP_006251375.1 | 1, 2 | 341 | |||
XM_008770307.1 | XP_008768529.1 | 1, 2 | 84 | |||
GenBank mRNA | CB757259.1 | - | 1, 2 | 77 | ||
FM044330.1 | - | 1, 2 | 51 | |||
FQ209875.1 | - | 1, 2 | 71 | |||
FQ223767.1 | - | 1, 2 | 38 |
