siRNA Details
| Gene Symbol: Urgcp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077660.2 | NP_001071128.1 | 6 | 2094 | ||
| XM_006251312.2 | XP_006251374.1 | 6 | 2555 | |||
| XM_006251313.2 | XP_006251375.1 | 6 | 2384 | |||
| XM_006251315.2 | XP_006251377.1 | 6 | 2187 | |||
| XM_008770307.1 | XP_008768529.1 | 5 | 2076 | |||
| GenBank mRNA | DN931704.1 | - | 1 | 588 | ||
| EX490902.1 | - | 1 | 160 | |||
| FQ209875.1 | - | 6 | 2114 | |||
| FQ222248.1 | - | 6 | 2086 | |||
| FQ223767.1 | - | 6 | 2081 | |||

Urgcp
Genomic Map