siRNA Details
| Gene Symbol: Aebp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100970.1 | NP_001094440.1 | 5, 6 | 1031 | ||
| XM_006251316.2 | XP_006251378.1 | 5, 6 | 1106 | |||
| GenBank mRNA | BC133065.1 | - | 5, 6 | 1031 | 7124847 | |

Aebp1
Genomic Map