siRNA Details
| Gene Symbol: Aebp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100970.1 | NP_001094440.1 | 18 | 2587 | ||
| XM_006251316.2 | XP_006251378.1 | 18 | 2659 | |||
| GenBank mRNA | BC133065.1 | - | 18 | 2587 | 7124847 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aebp1| Gene Symbol: Aebp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100970.1 | NP_001094440.1 | 18 | 2587 | ||
| XM_006251316.2 | XP_006251378.1 | 18 | 2659 | |||
| GenBank mRNA | BC133065.1 | - | 18 | 2587 | 7124847 | |