siRNA Details
| Gene Symbol: Adcy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107239.1 | NP_001100709.1 | 20 | 3004 | ||
| XM_008770320.1 | XP_008768542.1 | 22 | 3234 | |||
| XM_008770321.1 | XP_008768543.1 | 21 | 3162 | |||
| XM_008770322.1 | XP_008768544.1 | 20 | 2583 | |||

Adcy1
Genomic Map