siRNA Details
| Gene Symbol: Slc1a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198763.1 | NP_942058.1 | 3 | 591 | ||
| GenBank mRNA | AB103401.1 | - | 3 | 591 | ||
| FQ210448.1 | - | 819 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc1a4| Gene Symbol: Slc1a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198763.1 | NP_942058.1 | 3 | 591 | ||
| GenBank mRNA | AB103401.1 | - | 3 | 591 | ||
| FQ210448.1 | - | 819 | ||||