siRNA Details
Gene Symbol: Ehbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006251565.1 | XP_006251627.1 | 13, 14 | 2024 | ||
XM_006251566.1 | XP_006251628.1 | 12, 13 | 1919 | |||
XM_006251567.1 | XP_006251629.1 | 13, 14 | 2024 | |||
XM_006251568.1 | XP_006251630.1 | 11, 12 | 1796 | |||
XM_008770445.1 | XP_008768667.1 | 13, 14 | 1800 | |||
XM_008770446.1 | XP_008768668.1 | 12, 13 | 1919 | |||
XM_223664.7 | XP_223664.6 | 13, 14 | 2024 |
